Order Entry
Ireland
Orders LinkContactUsLinkComponent
(1S)-(-)-Borneol 97%

(1S)-(-)-Borneol 97%

Flammable
Irritant
(1S)-(-)-Borneol 97%
Certificates
(1S)-(-)-Borneol 97%

3 Options Available Below

Product Details & Documents
Synonyms:  (-)-Borneol, L(-)-Borneol, (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
  • CAS Number:464-45-9
  • Formula:C₁₀H₁₈O
  • MDL Number:MFCD00003759
  • Molecular Weight:154.25 g/mol
  • Merck Index:13,01328
  • Boiling Pt:210 °C (760 mmHg)
  • Melting Pt:204…208 °C
  • Density:1,011 g/cm³ (20 °C)
  • Flash Pt:65 °C
  • Storage Temperature:Ambient
  • EINECS:207-353-1
  • UN:1312
  • ADR:4.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    204 to 208 °C
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    -34.7° to -37.7° (20 °C, 589 nm) (c=5, C2H5OH)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 47,20 €
    47,20 €
    1 * 25 g
       47,20 €

  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 79,30 €
    79,30 €
    1 * 100 g
       79,30 €

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 334,00 €
    334,00 €
    1 * 500 g
       334,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.