Meglumine 99.0-101.0% (by acidimetry), EMPROVE® API ChP, Ph. Eur., JP, USP suitable for use as an active pharmaceutical ingredient low in endotoxins, SAFC®
: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
:6284-40-8
:
:H₃CNHCH₂(CH(OH))₄CH₂OH
:MFCD00004707
:210 °C (1013 hPa) (dec.)
:128…131 °C
:300 kg/m3
:195.22 g/mol
:12,06154
:Ambient
:228-506-9
- Environmentally Preferable:
Frequently Bought Together