
D-(-)-N-Methylglucamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6284-40-8
:
Formula:H₃CNHCH₂(CH(OH))₄CH₂OH
MDL Number:MFCD00004707
:195.22 g/mol
Merck Index:14,6078
Melting Pt:128…131 °C
EINECS:228-506-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together