L(+)-Serine, Millipore®
Catalog # 5650-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:105.09 g/mol
Merck Index:14,00846
Boiling Pt:259 °C (1013 hPa)
Melting Pt:215…225 °C
Density:1,6 g/cm³ (22 °C)
Storage Temperature:Ambient
EINECS:200-274-3
CAS Number:56-45-1
Formula:C₃H₇NO₃
MDL Number:MFCD00064224