L(+)-Serine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-45-1
:
Formula:C₃H₇NO₃
MDL Number:MFCD00064224
:105.09 g/mol
Merck Index:14,8460
Boiling Pt:259 °C (1013 hPa)
Melting Pt:214…224 °C (dec.)
Density:1,6 g/cm³ (22 °C)
Storage Temperature:Ambient
EINECS:200-274-3
Related Information
Frequently Bought Together