Order Entry
Ireland
ContactUsLinkComponent
Copper(II) sulphate 0.1 mol/l, Titripur® volumetric solution, Supelco®
Environment
Copper(II) sulphate 0.1 mol/l, Titripur® volumetric solution, Supelco®
Supplier:  Merck
Certificates
Environment
Copper(II) sulphate 0.1 mol/l, Titripur® volumetric solution, Supelco®
Supplier:  Merck
Certificates
Specification Test Results
  • Form:
    liquid
  • Amount-of-substance concentration:
    0.0995 - 0.1005
  • Measurement uncertainty:
    +/- 0.0003
  • Traceability:
    NIST SRM