
Hydrochloric acid 6 mol/l (6 N)
Catalog # HACH88449
Supplier: Hach
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
Density:1,06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324