Pesticide reference standard, Dieldrin 100 µg/ml in methanol
Catalog # ACCUP-037S
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:60-57-1
- Description:Dieldrin 100 µg/ml in methanol
- Formula:C₁₂H₈Cl₆O
- Description:Dieldrin