Order Entry
Ireland
ContactUsLinkComponent
Hydrochloric acid 1 mol/l (1 N)
Hydrochloric acid 1 mol/l (1 N)
Catalog # HACH2321353
Supplier:  Hach
CAS Number:  
Hydrochloric acid 1 mol/l (1 N)
Catalog # HACH2321353
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324