Order Entry
Ireland
ContactUsLinkComponent
Pesticide reference standard, 4,4'-TDE 100 µg/ml in methanol
Pesticide reference standard, 4,4'-TDE 100 µg/ml in methanol
Catalog # ACCUP-025S
Supplier:  AccuStandard
CAS Number:  
Pesticide reference standard, 4,4'-TDE 100 µg/ml in methanol
Catalog # ACCUP-025S
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    72-54-8
  • Description:
    4,4'-TDE 100 µg/ml in methanol
  • Formula:
    C₁₄H₁₀Cl₄
  • Description:
    4,4'-TDE

Specifications

About this item