Order Entry
Ireland
ContactUsLinkComponent
 
 
 
Sulphuric acid fuming (20 - 30% free SO₃), extra pure
 
undefined
 
 
Sulphuric acid fuming (20 - 30% free SO₃), extra pure
 

 

  • Appearance:
    Clear colorless to light brown Liquid
  • Assay:
    20 to 30 % (free SO3)
  • Residue after ignition:
    ≤0.01 %
  • Ammonium (NH4):
    ≤30 ppm
  • Arsenic (As):
    ≤2 ppm
  • Iron (Fe):
    ≤50 ppm

 

Frequently Bought Together