Order Entry
Ireland
Orders LinkContactUsLinkComponent
Rifampicin

Rifampicin

Glass bottle for solids, 5 g
Catalog # MOLE32609202-5G
Marketsource
Supplier:   Molekula
Rifampicin
Catalog # MOLE32609202-5G
Supplier:  Molekula
Marketsource
CAS Number:  13292-46-1
Rifampicin

Glass bottle for solids, 5 g

CAS Number:  13292-46-1
Price
255,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
  • CAS Number:13292-46-1
  • Formula:C₄₃H₅₈N₄O₁₂
  • MDL Number:MFCD00151389
  • Molecular Weight:822.95 g/mol
  • Merck Index:12,08382
  • Melting Pt:183…188 °C
  • EINECS:236-312-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.