Glyoxal 40% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-22-2
:
Formula:C₂H₂O₂
MDL Number:MFCD00006957
:58.04 g/mol
Merck Index:14,4509
Boiling Pt:104 °C
Melting Pt:–14 °C
Density:1,271
Storage Temperature:Ambient
EINECS:203-474-9
:GHS08
:GHS07
Related Information
Frequently Bought Together