Glyoxal 40% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-22-2
:
Formula:C₂H₂O₂
MDL Number:MFCD00006957
:58.04 g/mol
Melting Pt:–14 °C
Density:1,265 g/cm³
Flash Pt:>104 °C
EINECS:203-474-9
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to yellow Solution (May darken on storage)
- Titration excess NaOH:39 to 41 %
- Acidity (CH3COOH):≤0.25 %
Specification Test Results
Frequently Bought Together