Xanthan gum
Catalog # ROTH3557.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Density:1,5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:234-394-2
CAS Number:11138-66-2
MDL Number:MFCD00131256