
Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) for qualitative determination of reducing sugars
Catalog # ROTHN056.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Density:1,2 g/cm³ (20 °C)
UN:1824
ADR:8,II
:GHS05
:6381-59-5