Lactose monohydrate, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10039-26-6
:
Formula:C₁₂H₂₂O₁₁·1H₂O
:360.32 g/mol
Melting Pt:223 °C
Storage Temperature:Ambient
EINECS:200-559-2
Specification Test Results
- Spec. opt. rotation (calc. on anhydrous):54.4 - 55.9 °
- Ignition residue (SO4):Max. 0.1 %
- Loss on drying (120°C):4.0 - 5.5 %
- Cl (Chloride):Max. 100 ppm
- SO4 (Sulphate):Max. 100 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500)
Packaging Information
24945_BSE_LACTOSE 1H2O_REV0
BSE/TSE Certificates
spec_VWRC24945.291 spec_VWRC24945.360
Product specification
Frequently Bought Together