Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diaphragm cleaner with thiourea
Diaphragm cleaner with thiourea for pH-electrodes
Diaphragm cleaner with thiourea for pH-electrodes
83604.260
:
VWR Chemicals
:
Diaphragm cleaner with thiourea for pH-electrodes
83604.260
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
500 mL
Seq:
2725
Control:
Passes test
:
:
76.12 g/mol
:
177 °C
:
1,017 g/cm³ (20 °C)
:
Ambient
:
3082
:
9,III
:
GHS08
:
GHS07
:
GHS09