
Bromophenol blue 0,04% in ethanol, TECHNICAL
Catalog # 34307.237
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:669.97 g/mol
Storage Temperature:Ambient
EINECS:204-086-2
UN:1170
ADR:3,II
:GHS02
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875