
Thymol blue 0,04% in ethanol, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:76-61-9
:
Formula:C₂₇H₃₀O₅S
MDL Number:MFCD00005869
:466.6 g/mol
Storage Temperature:Ambient
EINECS:200-973-3
UN:1170
ADR:3,II
:GHS02
Specification Test Results
- Identification:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Glass bottle for liquids DIN 32 (ZPACK13415030)
Packaging Information
spec_VWRC34300.234
Product specification
VWR® Indicators
Product Literature
Frequently Bought Together