LITMUS 2% in aqueous solution, TECHNICAL pH-indicator
Catalog # 31725.265
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
EINECS:215-739-6
CAS Number:1393-92-6
MDL Number:MFCD00131528