Order Entry
Ireland
ContactUsLinkComponent
 
Toxic
Health
Environment
Nessler reagent for determination of ammonia and ammonium salts
Catalog # 31074.265
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Health
Environment
Nessler reagent for determination of ammonia and ammonium salts
Catalog # 31074.265
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Sensitivity:
    Passes test

Specification Test Results

About this item

 :
Boiling Pt:>100 °C (1013 hPa)
Density:1,2 g/cm³ (20 °C)
UN:2922
ADR:8 (6.1),II
 :GHS06
 :GHS08
 :GHS09
CAS Number:7783-33-7
MDL Number:MFCD00049656