


Ninhydrin 0,5% in 1-butanol, spray reagent for TLC
Catalog # 30960.226
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Aerosol can
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Suitable as reagent:Passes test
- Performance of spray:Passes test
Specification Test Results
About this item
:
:178.14 g/mol
Storage Temperature:Ambient
EINECS:207-618-1
UN:1950
ADR:2.1,
:GHS02
:GHS05
:GHS07
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791