Thiamine hydrochloride, TECHNICAL
Catalog # 28605.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:337.27 g/mol
Merck Index:13,09366
Melting Pt:252 °C
Density:1,401 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-641-8
CAS Number:67-03-8
Formula:C₁₂H₁₈Cl₂N₄OS
MDL Number:MFCD00012780