Formal saline, Gurr™ histological fixative
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
Storage Temperature:Ambient
EINECS:200-001-8
UN:3334
ADR:9,III
:GHS08
:GHS07
Specification Test Results
- pH (25°C):3.50 - 4.50
Specification Test Results
Related Information
Frequently Bought Together