Order Entry
Ireland
ContactUsLinkComponent
 
Fuchsin (acid), Gurr™ for microscopical staining
  VWRS34319.5000
 :  
Fuchsin (acid), Gurr™ for microscopical staining
  VWRS34319.5000
 :  

 

  • Pk:
    5 kg
  • Seq:
    4114

 

 

  • Dye content (titanometric):
    Min 60 %
  • Suitability for microscopy:
    Passes test
  • Loss on drying (110°C):
    Max 10 %
  • Absorptivity (A 1%/1cm, Lambda max., in dilute HCl):
    780 to 1310
  • Absorption max. Lambda (in dilute HCl):
    540 to 549 nm
  • TLC test:
    Passes test

 

 

 :
 :585.55 g/mol
 :12,00109
 :> 130 °C
 :Ambient
 :221-816-5
 :3244-88-0
 :C₂₀H₁₇N₃Na₂O₉S₃
 :MFCD00013286