Order Entry
Ireland
ContactUsLinkComponent
Vanadomolybdate reagent
Corrosive
Vanadomolybdate reagent
Catalog # 191844W
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Vanadomolybdate reagent
Catalog # 191844W
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Colour reaction:
    Satisfactory

Specification Test Results

About this item

 :
Boiling Pt:~100 °C (1013 hPa)
Density:1,2 g/cm³ (20 °C)
UN:2031
ADR:8,II
 :GHS05