Order Entry
Ireland
ContactUsLinkComponent
Florisil® 100-200 mesh for chromatography
Florisil® 100-200 mesh for chromatography
Catalog # 150263G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Florisil® 100-200 mesh for chromatography
Catalog # 150263G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Identification:
    Passes test
  • Particle size (100-200 US Mesh):
    Min. 70 %

Specification Test Results

About this item

 :
 :100.38 g/mol
EINECS:215-681-1
CAS Number:1343-88-0
Formula:MgO₃Si
MDL Number:MFCD00078249