



White spirit, 150…200 °C, SPECTRONORM® for atomic absorption spectroscopy
Catalog # 141976Y
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Density (20/4):0.750 - 0.790
- Al (Aluminium):Max. 0.02 ppm
- Ba (Barium):Max. 0.05 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.02 ppm
- Co (Cobalt):Max. 0.005 ppm
- Cr (Chromium):Max. 0.005 ppm
- Cu (Copper):Max. 0.2 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.02 ppm
- Mn (Manganese):Max. 0.005 ppm
- Mo (Molybdenum):Max. 0.005 ppm
- Na (Sodium):Max. 0.1 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.1 ppm
- Sr (Strontium):Max. 0.005 ppm
- Zn (Zinc):Max. 0.1 ppm
Specification Test Results
About this item
:
Boiling Pt:150…200 °C (1013 hPa)
Melting Pt:–15 °C
Density:0,77 g/cm³ (20 °C)
UN:3295
ADR:3,III
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:42 °C
EINECS:919-857-5
REACH:01-2119463258-33
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:64742-48-9
:101316-46-5