N,N-Dimethylacetamide ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:127-19-5
:
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686
:87.12 g/mol
Merck Index:13,03254
Boiling Pt:166,1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0,937 g/cm³ (20 °C)
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
UN:2810
ADR:6.1,III
REACH:01-2119459339-27
:GHS08
:GHS07
Specification Test Results
- Assay:≥ 99 %
- IR Spectrum:Passes test
- Density (20/4):0,940 - 0,.942
- Water:≤ 0.1 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85910.290 spec_VWRC85910.290 spec_VWRC85910.290 spec_VWRC85910.320 spec_VWRC85910.320
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together