Order Entry
Ireland
ContactUsLinkComponent
Masking tape, 101E
Masking tape, 101E
Catalog # MMMA1013650
Supplier:  3M
Masking tape, 101E
Catalog # MMMA1013650
Supplier:  3M
Specifications
  • Width:
    36 mm
  • Colour:
    Beige
  • Description:
    Masking tape, rubber adhesive
  • Pk:
    24 Roll
Specifications
About this item
3M™ Masking Tape 101E is a smooth, multi-purpose crepe paper tape for room temperature indoor light-duty applications.
  • Good initial tack to a wide variety of surfaces in different applications
  • Designed for indoor use in light-duty applications
  • Ideal for cropping, temporary mooring, packaging, and tying
  • Easy to write on for labelling
The cost-effective general-purpose masking tape is a chamois-colored, 0,12 mm thick tape with rubber adhesive for general applications. It provides conformability on irregular surfaces and around corners, and is hand-tearable for easy use. Ideal for non-critical paint masking, but also bundling, sealing, attaching, and other applications.
Length: 50,0 m.