Order Entry
Ireland
ContactUsLinkComponent
n-Tetradecane ≥99.0%, GPR RECTAPUR®
Health
Irritant
n-Tetradecane ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
Irritant
n-Tetradecane ≥99.0%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99,0%
  • Identification (IR):
    Passes test
  • Density (25/4):
    0,761 - 0,765

Specification Test Results

Frequently Bought Together