Human Recombinant FGF-10 (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Fibroblast growth factor 10 (FGF-10) is a growth factor that is important during embryonic development, especially during lung, limb, brain, heart, and kidney morphogenesis.FGF-10 is expressed in mesenchymal cells and facilitates epithelial-mesenchymal signaling through binding the epithelially expressed FGF receptor 2b (FGFR2b). FGF-10 also functions as a mitogen for keratinising epidermal cells, and induces the migration and invasion of cancer cells.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤200 ng/ml, Bioactivity ≥5,0×10³ units/mg
- Gene ID:O15520
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Keratinocyte growth factor 2
- Protein/peptide name:FGF-10 (AF)
- Purity:≥95%
- Molecular weight:19,3 kDa
- Sequence:MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEI GVVAVKAINS NYYLAMNKKG KLYGSKEFNNDCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together