Order Entry
Ireland
ContactUsLinkComponent
Mouse Recombinant TNF-A (from E. coli)
Mouse Recombinant TNF-A (from E. coli)
Certificates
Mouse Recombinant TNF-A (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity Assay: Cytolysis of mouse L929 cells in the presence of Actinomycin D, ED₅₀ ≤0,1 ng/ml, Bioactivity ≥1,0×10⁷ units/mg
  • Gene ID:
    P06804
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Cytotoxin|DIF|Necrosin|Cachexin|TNFSF2|TNF|Cachectin
  • Protein/peptide name:
    TNF-A (AF)
  • Purity:
    ≥95%
  • Molecular weight:
    17,4 kDa
  • Sequence:
    MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together