STERI-PEROX®, Cleaning Agent
Supplier: VELTEK ASSOCIATES
Certificates
About this item
Sterile, ready-to-use hydrogen peroxide solution for pharmaceutical and biotechnology operations. Formulated with USP Water for Injection
- Filled in an ISO 5 (Grade A/B, Former Class 100)
- Prefiltered to 0,2 um
- Completely traceable and validated for sterility and shelf life
- Avaliable in 3% and 6% concentrations
- 16 oz and 32 oz containers come with sterile spray attachments
- Low residue content
STERI-PEROX reduces exposure concerns for VOC’s in cleanroom operations, leaves a low remaining residue, and is designed for most washable, non-porous, hard, inanimate environmental surfaces.
Certifications: Delivered with lot specific Certificate of Analysis and Certificate of Sterility.
Packaging: The products are packaged to comply with the requirements for transportation into a classified area. Each container is double bagged packaged sterile.
Frequently Bought Together