
Lauric acid ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:143-07-7
:
Formula:C₁₂H₂₄O₂
MDL Number:MFCD00002736
:200.32 g/mol
Merck Index:13,05400
Boiling Pt:296 °C (1013 hPa)
Melting Pt:42…45 °C
Density:0,875 g/cm³ (20 °C)
Flash Pt:>150 °C
EINECS:205-582-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99 %
- Melting point:43 --> 45 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC87071.180 spec_VWRC87071.290 spec_VWRC87071.460
Product specification
Frequently Bought Together