Order Entry
Ireland
ContactUsLinkComponent
 
Sodium bromate ≥99%, GPR RECTAPUR®
 
undefined
Sodium bromate ≥99%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 99 %
  • pH (5 %):
    5 --> 9
  • Br (Bromide):
    Max. 0.05 %
  • SO4 (Sulphate):
    Max. 0.05 %

 

Frequently Bought Together