NFKB p65 (rel A) (phospho-Ser529) control peptide
Catalog # ROCK000-000-266
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Pk:50 µG
- Seq:02
- Protein/peptide name:NFKB p65 (rel A) (phospho-Ser529) Control Peptide
- Tested applications:Control Peptide