Order Entry
Ireland
ContactUsLinkComponent
 
Irritant
Isonicotinic acid, GPR RECTAPUR®
Catalog # 87093.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Isonicotinic acid, GPR RECTAPUR®
Catalog # 87093.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :123.11 g/mol
Merck Index:13,05206
Boiling Pt:396 °C (1013 hPa)
Melting Pt:315…319 °C
Storage Temperature:Ambient
EINECS:200-228-2
 :GHS07
CAS Number:55-22-1
Formula:C₆H₅NO₂
MDL Number:MFCD00006429