


Salicylic acid 99.5-100.5% (dried basis), meets analytical specification of BP, Ph. Eur., USP, Fluka™
Catalog # HONC27301H-1KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- assay (calc. to the dried substance):99.5 - 100.5 %
- assay (HPLC, ex dried substance):98.0 - 102.0 %
- melting range:158 - 161 °C
- loss on drying:Max. 0.3 %
- water (Karl Fischer):Max. 0.2 %
- sulfated ash:Max. 0.05 %
- heavy metals (as Pb):Max. 0.001 %
- chloride (Cl):Max. 0.005 %
- sulfate (SO4):Max. 0.02 %
- identity:complying
- appearance of the solution:complying
- related subst. (HPLC):Max. 0.2 %
- Residual solvents:complying
Specification Test Results
About this item
:
:138.12 g/mol
Boiling Pt:211 °C
Density:1,440 g/cm3
:GHS07
:GHS05
:GHS08
CAS Number:69-72-7
Formula:(HO)C₆H₄CO₂H
MDL Number:MFCD00002439