General purpose gloves, Showa 341
Supplier: Showa
Certificates
About this item
These nylon/polyester knitted gloves are hard-wearing and highly tactile, even in wet environments.
- Natural rubber coating on the palm is water-resistant
- Rough surface grip enables optimum dexterity when working with small components or heavy components
- Elasticated wrist cuff prevents dirt and debris from entering the glove
- Seamless knit offers ultimate comfort during long periods of wear
Typical applications include: Decoration, drainage, piping, flashings and seals, guttering and drain pipe, electrical wiring, insulation, roofing application membranes.
Certifications: EN 388:2016 (2121X); Cat. II
Packaging: 10 pairs per polybag.
Specifications
- Catalog No:
- 112-6006
- 112-6005
- 112-6004
- 112-6003
- Model:
- Showa 341
- Showa 341
- Showa 341
- Showa 341
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon/polyester
- Nylon/polyester
- Nylon/polyester
- Nylon/polyester
- Finish:
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)