Order Entry
Ireland
ContactUsLinkComponent
General purpose gloves, Showa 370
General purpose gloves, Showa 370
Supplier:  Showa
Certificates
General purpose gloves, Showa 370
Supplier:  Showa
Specifications
  • Catalog No:
  • 112-5796
  • 112-5795
  • 112-5794
  • 112-5793
  • 112-5792
  • Model:
  • Showa 370
  • Showa 370
  • Showa 370
  • Showa 370
  • Showa 370
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Finish:
  • Nitrile coating standard finish
  • Nitrile coating standard finish
  • Nitrile coating standard finish
  • Nitrile coating standard finish
  • Nitrile coating standard finish
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)
  • EN 388:2016 (4121X)