ESD safety shoes, Boa® Fit System, uvex 1, 6566
Supplier: UVEX
Certificates
About this item
Versatile compact design S2 SRC safety shoe with wide fit helps in reducing the strain experienced by the body during the working day and wearer comfort is optimised.
- Breathable, perforated microvelours
- Soft padding on collar and tongue
- Boa® Fit System for individual adjustment and optimum fit all day long
- Meets the ESD specification with volume resistance less than 35 megaohms
- Removable anti-static comfort insole with moisture control system and additional shock absorption
Safety shoes features virtually seam-free construction to eliminate pressure points, as well as an ergonomically designed outsole that offers excellent slip resistance. 100% metal-free uvex xenova® toe cap gives anatomical shape, good lateral stability, no thermal conductivity. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/yellow
Certifications: EN 61340 (ESD); EN ISO 20345:2011, S2, SRC
Frequently Bought Together