Order Entry
Ireland
ContactUsLinkComponent
Sodium sulphate decahydrate ≥99.0% (by titrimetric analysis on dry substance), crystallised, Puriss. p.a., Fluka™

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • pH (5 %, 20°C):
    5.0 - 8.0
  • Appearance of the solution:
    complying
  • Loss on drying (130°C):
    53 - 57 %
  • Calcium (Ca):
    Max. 20 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 20 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Phosphate (PO₄):
    Max. 10 ppm
  • Total N:
    Max. 5 ppm

Specification Test Results

About this item

 :
 :322.2 g/mol
Merck Index:13,08755
Melting Pt:32,4 °C
Density:1,46 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7727-73-3
Formula:Na₂SO₄·10H₂O
MDL Number:MFCD00149189