Order Entry
Ireland
ContactUsLinkComponent
Acetonitrile with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Acetonitrile with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Catalog # 14272-2L
CAS Number:  
Acetonitrile with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Catalog # 14272-2L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

Specification Test Results

  • Formic acid:
    0.095 - 0.105 Vol.%
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Aluminium (Al):
    Max. 0.03 ppm
  • Calcium (Ca):
    Max. 0.1 ppm
  • Iron (Fe):
    Max. 0.05 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.03 ppm
  • Sodium (Na):
    Max. 0.1 ppm
  • Transmittance at 254 nm:
    Min. 90 %
  • Transmittance at 230 nm:
    Min. 15 %
  • Transmittance at 210 nm:
    Min. 5 %
  • Suitability for the UHPLC-MS:
    complying
  • UHPLC Gradient drift 254 nm:
    Max. 30 mAU
  • UHPLC Gradient peak 254 nm:
    Max. 2 mAU
  • UHPLC-MS ESI(+) (Reserpine):
    Max. 20 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb
  • Fluorescence (chinin) at 254 nm:
    Max. 0.5 ppb

Specification Test Results

About this item

 :
Boiling Pt:81 °C
Melting Pt:–46 °C
Density:0,78 g/cm³ (20 °C)
Flash Pt:5 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :75-05-8
MDL Number:MFCD00001878