N,N-Dimethylglycine ≥99% (by total nitrogen and titration analysis), LiChropur™ derivatisation reagent, for HPLC, for LC-MS, Supelco®
Catalog # 05022-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:103.12 g/mol
CAS Number:1118-68-9
Formula:C₄H₉NO₂
MDL Number:MFCD00004283