Order Entry
Ireland
ContactUsLinkComponent
1-Hexanol ≥98.0% (by GC), GPR RECTAPUR®
Irritant
1-Hexanol ≥98.0% (by GC), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
1-Hexanol ≥98.0% (by GC), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Assay (GC):
    min. 98.0 %
  • Density (d20°C/ 4°C):
    0.817 - 0.821
  • Identity (IR):
    Passes test

Specification Test Results

Frequently Bought Together