Order Entry
Ireland
ContactUsLinkComponent
1,3-Dimethyl-2-imidazolidinone ≥99%, GPR RECTAPUR®
1,3-Dimethyl-2-imidazolidinone ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
1,3-Dimethyl-2-imidazolidinone ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %

Specification Test Results

Frequently Bought Together