Pyrogallol red indicator for metal titration
Catalog # 1.07534.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance of solution:conforms
- Loss on drying (110 °C):≤ 5.0 %
- Suitability as indicator (for metal titration):conforms
Specification Test Results
About this item
:
:400.36 g/mol
Melting Pt:>300 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:32638-88-3
Formula:C₁₉H₁₂O₈S
MDL Number:MFCD00005046