1,3-Dinitroglycerin solution, certified reference material, Cerilliant®, Supelco®
Catalog # D-011-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:623-87-0
- Description:1,3-Dinitroglycerin solution
- Formula:C₃H₆N₂O₇
- Description:1,3-Dinitroglycerin solution
- Pk Info:Ampoule of 1 ml
Specifications
About this item
1,3-Dinitroglycerin is a major plasma metabolite of the pharmaceutical vasodilator nitroglycerin.
This Certified Spiking Solution® is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology. Nitroglycerin is used to treat heart conditions such as angina and chronic heart failure.
Packaging: Snap-N-Spike®/Snap-N-Shoot®